Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100752154 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507016.2 | XP_003507064.2 | 5 - 6 | 851 | 105 | |
XM_007647547.1 | XP_007645737.1 | 6 - 7 | 999 | 105 | ||
XM_007647548.1 | XP_007645738.1 | 4 - 5 | 766 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |