Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497857.1 | XP_003497905.1 | 33 - 34 | 5329 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |