Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007640044.1 | XP_007638234.1 | 1 - 2 | 404 | 102 | |
XM_007640045.1 | XP_007638235.1 | 1 - 2 | 404 | 102 | ||
GenBank mRNA | JI874387.1 | - | 1 - 2 | 396 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |