Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Folr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506496.1 | XP_003506544.1 | 2 - 3 | 331 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Transporter Probe spans exons |