Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Surf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003511729.2 | XP_003511777.1 | 1 - 2 | 176 | 102 | |
GenBank mRNA | JI883990.1 | - | 1 - 2 | 163 | 102 | |
JP055378.1 | - | 1 - 2 | 163 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |