Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100774007 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503446.2 | XP_003503494.1 | 3 - 4 | 689 | 78 | |
GenBank mRNA | JP064935.1 | - | 1 - 2 | 115 | 78 |