Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007640818.1 | XP_007639008.1 | 1 - 2 | 174 | 82 | |
GenBank mRNA | JP056480.1 | - | 1 - 2 | 174 | 82 |