Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il9r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007652360.1 | XP_007650550.1 | 6 - 7 | 840 | 72 | |
XM_007652362.1 | XP_007650552.1 | 6 - 7 | 837 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Apoptosis Detect Genomic DNA Probe spans exons |