Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc39a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501629.2 | XP_003501677.1 | 2 - 3 | 587 | 79 | |
XM_007642290.1 | XP_007640480.1 | 3 - 4 | 496 | 79 | ||
XM_007642291.1 | XP_007640481.1 | 3 - 4 | 787 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Transporter 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |