Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003505971.2 | XP_003506019.1 | 3 - 4 | 326 | 76 | |
GenBank mRNA | JI894452.1 | - | 3 - 4 | 320 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |