Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lin28b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509668.1 | XP_003509716.1 | 3 - 4 | 479 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |