Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcmo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007609082.1 | XP_007607272.1 | 2 - 3 | 96 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |