Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppm1n | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007645954.1 | XP_007644144.1 | 1 - 2 | 294 | 103 | |
XM_007645955.1 | XP_007644145.1 | 1 - 2 | 294 | 103 | ||
XM_007645956.1 | XP_007644146.1 | 1 - 2 | 294 | 103 | ||
XM_007645957.1 | XP_007644147.1 | 1 - 2 | 294 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Phosphatase 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |