Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Etnk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007633623.1 | XP_007631813.1 | 6 - 7 | 849 | 88 | |
GenBank mRNA | JI874837.1 | - | 2 - 3 | 276 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |