Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhoc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003512964.2 | XP_003513012.1 | 3 - 4 | 463 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Probe spans exons |