Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mia2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514701.1 | XP_003514749.1 | 3 - 4 | 330 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |