Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scnm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498667.2 | XP_003498715.1 | 3 - 4 | 327 | 95 | |
GenBank mRNA | JI863698.1 | - | 3 - 4 | 327 | 95 | |
JP057693.1 | - | 3 - 4 | 327 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |