Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mefv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007649324.1 | XP_007647514.1 | 6 - 7 | 1878 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |