Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: L1td1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007614762.1 | XP_007612952.1 | 1 - 2 | 896 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |