Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nedd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502054.2 | XP_003502102.1 | 3 - 4 | 440 | 66 | |
GenBank mRNA | JI886815.1 | - | 3 - 4 | 442 | 66 | |
JP062071.1 | - | 3 - 4 | 442 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |