Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spty2d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509345.2 | XP_003509393.1 | 3 - 4 | 1902 | 88 | |
XM_007649930.1 | XP_007648120.1 | 4 - 5 | 1845 | 88 | ||
GenBank mRNA | JI875009.1 | - | 1 - 2 | 354 | 88 | |
JP065245.1 | - | 1 - 2 | 354 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |