Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499311.2 | XP_003499359.2 | 2 - 3 | 1937 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Probe spans exons |