Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007654255.1 | XP_007652445.1 | 9 - 10 | 1547 | 68 | |
XM_007654258.1 | XP_007652448.1 | 9 - 10 | 1548 | 68 | ||
XM_007654261.1 | XP_007652451.1 | 8 - 9 | 1389 | 68 | ||
XM_007654267.1 | XP_007652457.1 | 9 - 10 | 1547 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |