Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007654255.1 | XP_007652445.1 | 12 - 13 | 2023 | 61 | |
XM_007654261.1 | XP_007652451.1 | 11 - 12 | 1865 | 61 | ||
XM_007654267.1 | XP_007652457.1 | 12 - 13 | 2023 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |