Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm2d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497604.2 | XP_003497652.1 | 2 - 3 | 279 | 113 | |
GenBank mRNA | JI862682.1 | - | 1 - 2 | 81 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |