Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxcl16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007608718.1 | XP_007606908.1 | 1 - 2 | 171 | 67 | |
GenBank mRNA | JI888463.1 | - | 1 - 2 | 171 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Probe spans exons |