Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003512856.2 | XP_003512904.1 | 2 - 3 | 258 | 65 | |
GenBank mRNA | JI893724.1 | - | 2 - 3 | 275 | 65 | |
JP042442.1 | - | 2 - 3 | 275 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Probe spans exons |