Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510044.2 | XP_003510092.1 | 3 - 4 | 325 | 87 | |
GenBank mRNA | JI896010.1 | - | 3 - 4 | 395 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |