Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100757150 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007649867.1 | XP_007648057.1 | 3 - 4 | 758 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |