Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab3il1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003513179.2 | XP_003513227.1 | 9 - 10 | 1160 | 66 | |
XM_007653498.1 | XP_007651688.1 | 8 - 9 | 941 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |