Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnni1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007634464.1 | XP_007632654.1 | 9 - 10 | 783 | 55 | |
XM_007634468.1 | XP_007632658.1 | 9 - 10 | 756 | 55 | ||
XM_007634477.1 | XP_007632667.1 | 6 - 7 | 771 | 55 | ||
XM_007634480.1 | XP_007632670.1 | 8 - 9 | 693 | 55 | ||
XM_007634485.1 | XP_007632675.1 | 10 - 11 | 769 | 55 | ||
XM_007634493.1 | XP_007632683.1 | 8 - 9 | 682 | 55 | ||
XM_007634502.1 | XP_007632692.1 | 8 - 9 | 649 | 55 | ||
XM_007634509.1 | XP_007632699.1 | 8 - 9 | 661 | 55 | ||
XM_007634516.1 | XP_007632706.1 | 8 - 9 | 654 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Probe spans exons |