Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Leprel4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504621.2 | XP_003504669.1 | 4 - 5 | 601 | 96 | |
GenBank mRNA | JI876767.1 | - | 3 - 4 | 343 | 96 | |
JP047664.1 | - | 3 - 4 | 343 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |