Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jph4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007642692.1 | XP_007640882.1 | 3 - 4 | 966 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |