Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h12c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007643376.1 | XP_007641566.1 | 2 - 3 | 1120 | 106 | |
GenBank mRNA | JI876926.1 | - | 2 - 3 | 328 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |