Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rxra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007653390.1 | XP_007651580.1 | 3 - 4 | 599 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Nuclear Receptor Probe spans exons |