Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmga2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007607994.1 | XP_007606184.1 | 1 - 2 | 365 | 72 | |
XM_007607997.1 | XP_007606187.1 | 1 - 2 | 128 | 72 | ||
XM_007608001.1 | XP_007606191.1 | 1 - 2 | 364 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Probe spans exons |