Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhcr7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003505694.2 | XP_003505742.1 | 3 - 4 | 341 | 96 | |
XM_007646172.1 | XP_007644362.1 | 4 - 5 | 596 | 96 | ||
XM_007646173.1 | XP_007644363.1 | 5 - 6 | 642 | 96 | ||
XM_007646174.1 | XP_007644364.1 | 3 - 4 | 484 | 96 | ||
XM_007646175.1 | XP_007644365.1 | 4 - 5 | 616 | 96 | ||
GenBank mRNA | JI880296.1 | - | 2 - 3 | 316 | 96 | |
JP064895.1 | - | 2 - 3 | 317 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |