Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003515456.1 | XP_003515504.1 | 3 - 4 | 650 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter Detect Genomic DNA Probe spans exons |