Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003515456.1 | XP_003515504.1 | 7 - 8 | 1274 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Transporter Detect Genomic DNA Probe spans exons |