Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc42ep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510269.2 | XP_003510317.1 | 1 - 2 | 819 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |