Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Haus3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003508850.2 | XP_003508898.1 | 2 - 3 | 1155 | 100 | |
XM_007649465.1 | XP_007647655.1 | 2 - 3 | 1612 | 100 | ||
XM_007649466.1 | XP_007647656.1 | 3 - 4 | 1209 | 100 | ||
GenBank mRNA | JI870179.1 | - | 1 - 2 | 574 | 100 | |
JP043833.1 | - | 1 - 2 | 574 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Probe spans exons |