Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504306.2 | XP_003504354.1 | 3 - 4 | 348 | 73 | |
XM_007644839.1 | XP_007643029.1 | 3 - 4 | 329 | 73 | ||
XM_007644840.1 | XP_007643030.1 | 3 - 4 | 302 | 73 | ||
XM_007644841.1 | XP_007643031.1 | 3 - 4 | 348 | 73 | ||
GenBank mRNA | JI893296.1 | - | 1 - 2 | 96 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |