Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arsi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007646834.1 | XP_007645024.1 | 1 - 2 | 121 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |