Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdzd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003511555.2 | XP_003511603.1 | 6 - 7 | 659 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Probe spans exons |