Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007649253.1 | XP_007647443.1 | 1 - 2 | 86 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |