Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504581.2 | XP_003504629.1 | 4 - 5 | 502 | 91 | |
GenBank mRNA | JI865108.1 | - | 4 - 5 | 480 | 91 | |
JP062945.1 | - | 4 - 5 | 484 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |