Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003508435.2 | XP_003508483.1 | 2 - 3 | 741 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |