Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdhd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007641739.1 | XP_007639929.1 | 6 - 7 | 753 | 96 | |
XM_007641740.1 | XP_007639930.1 | 6 - 7 | 728 | 96 | ||
XM_007641741.1 | XP_007639931.1 | 6 - 7 | 707 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |