Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: R3hdml | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003508917.2 | XP_003508965.1 | 3 - 4 | 515 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Protease Probe spans exons |