Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100758463 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502409.2 | XP_003502457.1 | 4 - 5 | 531 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |